Assay Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173364.5 | NP_775540.4 | 7 | 5197 | 69 | |
| GenBank mRNA | AK032835.1 | - | 1 | 1391 | 69 | |
| AK035697.1 | - | 1 | 1553 | 69 | ||
| BC099896.1 | - | 6 | 5084 | 30545532 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
T cell activation inhibitor, mitochondrial |
Tcaim |
D9Ertd402e, Gm1129, TOAG-1 |
-13.3kb | Anti-sense | Chr. 9 - 122805361 - 122836332 on NCBI Build GRCm38 |
| Downstream | zinc finger with KRAB and SCAN domains 7 | Zkscan7 |
BC042569, Zfp167 |
35.5kb | Anti-sense | Chr. 9 - 122886979 - 122898619 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp445
Genomic Map