Assay Details
| Gene Symbol: A930009E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035831.1 | - | 1 | 2148 | 97 | |
| BC051426.1 | - | 1 | 3836 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 178B | Tmem178b |
EG434008, Gm5567 |
-3.4kb | Sense | Chr. 6 - 39872540 - 40248353 on NCBI Build GRCm38 |
| Downstream | acylglycerol kinase | Agk |
2610037M15Rik, 6720408I04Rik, AI465370, Mulk |
70.0kb | Sense | Chr. 6 - 40325155 - 40397562 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930009E05Rik
Genomic Map