Assay Details
| Gene Symbol: 9630017O17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035916.1 | - | 1 | 954 | 100 | |
| AK160690.1 | - | 1 | 936 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | asteroid homolog 1 (Drosophila) | Aste1 |
1100001A21Rik, AV006403 |
-849.0 | Sense | Chr. 9 - 105395294 - 105408298 on NCBI Build GRCm38 |
| Downstream |
ATPase, Ca++-sequestering |
Atp2c1 |
1700121J11Rik, ATP2C1A, AW061228, BCPM, D930003G21Rik, HHD, SPCA, pmr1 |
93.0 | Anti-sense | Chr. 9 - 105411362 - 105521257 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9630017O17
Genomic Map