Assay Details
| Gene Symbol: 9530082P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037113.1 | - | 464 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | progestin and adipoQ receptor family member IV | Paqr4 | 1500004C10Rik | -4.9kb | Anti-sense | Chr. 17 - 23736186 - 23740898 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

9530082P21Rik
Genomic Map