Assay Details
| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037324.1 | - | 1 | 1449 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Kruppel-like factor 7 (ubiquitous) | Klf7 | 9830124P08Rik | -454.0kb | Anti-sense | Chr. 1 - 64033727 - 64122341 on NCBI Build GRCm38 |
| Downstream | methyltransferase like 21A | Mettl21a |
2310038H17Rik, AI464204, Fam119a |
27.3kb | Anti-sense | Chr. 1 - 64606473 - 64620421 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Creb1
Genomic Map