Assay Details
| Gene Symbol: A330043J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039443.1 | - | 1 | 2555 | 83 | |
| AK039581.1 | - | 1 | 2555 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA binding motif protein 15 | Rbm15 |
C230088J01Rik, mKIAA1438 |
-107.1kb | Sense | Chr. 3 - 107326110 - 107333289 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 6 (neurotransmitter transporter), member 17 |
Slc6a17 |
D130012J15Rik, Ntt4 |
24.6kb | Sense | Chr. 3 - 107467548 - 107518018 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A330043J11Rik
Genomic Map