Assay Details
| Gene Symbol: Snhg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039454.1 | - | 752 | 79 | ||
| AK080843.1 | - | 1243 | 79 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 116-like 1 |
Snord116l1 | Pwcr1l1 | -79.4kb | Sense | Chr. 7 - 59894622 - 59894711 on NCBI Build GRCm38 |
| Downstream |
small nucleolar RNA, C/D box 64 |
Snord64 | MBII-13 | 4.1kb | Sense | Chr. 7 - 59978811 - 59978856 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snhg14
Genomic Map