Assay Details
| Gene Symbol: 5830428M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038060.1 | 1 - 2 | 68 | 98 | ||
| GenBank mRNA | AK039829.1 | - | 1 - 2 | 65 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP-ribosylation factor 6 | Arf6 |
AI788669, AW496366 |
-98.5kb | Anti-sense | Chr. 12 - 69372150 - 69375980 on NCBI Build GRCm38 |
| Downstream | valosin containing protein lysine (K) methyltransferase | Vcpkmt |
Gm71, Mettl21d, Mettl221d, VCP-KMT |
101.2kb | Sense | Chr. 12 - 69576730 - 69583109 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

5830428M24Rik
Genomic Map