Assay Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271768.1 | NP_001258697.1 | 5 | 2802 | 76 | |
| NM_024469.2 | NP_077789.1 | 5 | 2673 | 76 | ||
| GenBank mRNA | AK032333.1 | - | 1 | 483 | 76 | |
| AK040945.1 | - | 1 | 453 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras association (RalGDS/AF-6) domain family (N-terminal) member 8 | Rassf8 |
5133400D11Rik, AA815821, AW123240, Hoj1, mHoj-1 |
-40.1kb | Anti-sense | Chr. 6 - 145746562 - 145821079 on NCBI Build GRCm38 |
| Downstream | sarcospan | Sspn | Krag | 69.1kb | Anti-sense | Chr. 6 - 145931150 - 145965225 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bhlhe41
Genomic Map