Assay Details
| Gene Symbol: Gm6416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046023.1 | 2 - 3 | 148 | 69 | ||
| GenBank mRNA | AK041110.1 | - | 2 - 3 | 151 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
poly (ADP-ribose) polymerase family, member 8 |
Parp8 |
2810430O08Rik, ARTD16, D13Ertd275e |
-104.2kb | Anti-sense | Chr. 13 - 116850842 - 117025913 on NCBI Build GRCm38 |
| Downstream | embigin | Emb |
AL022799, Gp70 |
90.1kb | Sense | Chr. 13 - 117220573 - 117274415 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6416
Genomic Map