Assay Details
| Gene Symbol: Gm4668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041339.1 | - | 1 - 2 | 217 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transforming growth factor, beta receptor II |
Tgfbr2 |
1110020H15Rik, AU042018, DNIIR, RIIDN, TBR-II, TbetaR-II, TbetaRII |
-13.8kb | Anti-sense | Chr. 9 - 116087695 - 116175363 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA D730003K21 gene | D730003K21Rik | 136.3kb | Sense | Chr. 9 - 116327308 - 116339659 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm4668
Genomic Map