Assay Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042209.1 | - | 1 | 1150 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 331 | Mir331 |
Mirn331, mmu-mir-331 |
Sense | Chr. 10 - 93963768 - 93963863 on NCBI Build GRCm38 | |
| Downstream |
FYVE, RhoGEF and PH domain containing 6 |
Fgd6 |
AA123052, Etohd4, ZFYVE24 |
72.2kb | Anti-sense | Chr. 10 - 94036001 - 94145339 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vezt
Genomic Map