Assay Details
| Gene Symbol: Sh2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011364.3 | NP_035494.1 | 2 - 3 | 317 | 98 | |
| GenBank mRNA | AF072903.1 | - | 2 - 3 | 364 | 98 | |
| AF097632.1 | - | 2 - 3 | 263 | 98 | ||
| AF097633.1 | - | 2 - 3 | 256 | 98 | ||
| AK040076.1 | - | 2 - 3 | 318 | 98 | ||
| AK042476.1 | - | 146 | 98 | |||
| BC065390.1 | - | 2 - 3 | 287 | 98 | ||
| BC147367.1 | - | 2 - 3 | 227 | 9056424 | 98 | |
| BC147368.1 | - | 2 - 3 | 227 | 9056435 | 98 | |
| BC147807.1 | - | 2 - 3 | 90 | 9087524 | 98 | |
| BC147811.1 | - | 2 - 3 | 90 | 9087528 | 98 | |
| KF811030.1 | - | 3 - 4 | 256 | 98 | ||
More Information
Set Membership: |
|

Sh2d1a
Genomic Map