Assay Details
| Gene Symbol: Klhl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042526.1 | - | 207 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WDYHV motif containing 1 | Wdyhv1 |
2410187C16Rik, AU014961, AW550036, Ntaq1 |
-55.8kb | Sense | Chr. 15 - 58141334 - 58158654 on NCBI Build GRCm38 |
| Downstream | annexin A13 | Anxa13 |
1810034H17Rik, AV055219 |
20.6kb | Anti-sense | Chr. 15 - 58341464 - 58389274 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Klhl38
Genomic Map