Assay Details
| Gene Symbol: Gm4430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042594.1 | - | 1852 | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | expressed sequence AI847159 | AI847159 | OTTMUSG00000015420 | -16.6kb | Sense | Chr. 2 - 129178148 - 129180673 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 20, member 1 |
Slc20a1 |
AI607883, Glvr-1, Glvr1 |
877.0 | Anti-sense | Chr. 2 - 129198773 - 129211615 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm4430
Genomic Map