Assay Details
| Gene Symbol: 9330133O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045696.1 | 1 | 83 | 101 | ||
| GenBank mRNA | AK042662.1 | - | 1 | 48 | 101 | |
| AK049744.1 | - | 1 | 84 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mevalonate (diphospho) decarboxylase | Mvd |
C78718, Mpd |
-270.0 | Anti-sense | Chr. 8 - 122433596 - 122443422 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 20735 |
Gm20735 | 197.0 | Anti-sense | Chr. 8 - 122445268 - 122451322 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049744.1 transcripts.
Set Membership: |
|

9330133O14Rik
Genomic Map