Assay Details
| Gene Symbol: F730043M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015602.2 | 3 - 4 | 829 | 98 | ||
| GenBank mRNA | AK043182.1 | - | 3 - 4 | 830 | 98 | |
| AK162927.1 | - | 3 - 4 | 775 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cadherin-related family member 3 | Cdhr3 |
1110049B09Rik, BB172694, Cdh28 |
-42.9kb | Sense | Chr. 12 - 33033796 - 33093037 on NCBI Build GRCm38 |
| Downstream | ataxin 7-like 1 | Atxn7l1 |
2810423G08Rik, Atxn7l4 |
7.9kb | Anti-sense | Chr. 12 - 33147639 - 33376408 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

F730043M19Rik
Genomic Map