Assay Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036940.1 | - | 1 | 1600 | 68 | |
AK043267.1 | - | 1 | 1446 | 68 | ||
AK172512.1 | - | 1 | 1542 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 3961 | Mir3961 | mmu-mir-3961 | -826.6kb | Anti-sense | Chr. 13 - 82698275 - 82698333 on NCBI Build GRCm38 |
Downstream |
predicted gene, 26803 |
Gm26803 | 139.6kb | Anti-sense | Chr. 13 - 83666358 - 83730230 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
