Assay Details
| Gene Symbol: Gm14317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043662.1 | - | 1 - 2 | 268 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylinositol glycan anchor biosynthesis, class T |
Pigt |
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7 |
-42.2kb | Sense | Chr. 2 - 164497511 - 164508301 on NCBI Build GRCm38 |
| Downstream | WAP four-disulfide core domain 2 | Wfdc2 |
1600023A02Rik, HE4, WAP5 |
4.3kb | Sense | Chr. 2 - 164562563 - 164568510 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm14317
Genomic Map