Assay Details
| Gene Symbol: Rab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043699.1 | - | 1 | 4052 | 65 | |
| AK169946.1 | - | 1 | 4033 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | centriolin | Cntrl |
6720467O09Rik, AA968343, Cep1, Cep110, IB3/5, Ma2a8, b2b1468Clo |
-17.8kb | Anti-sense | Chr. 2 - 35109492 - 35178822 on NCBI Build GRCm38 |
| Downstream | predicted gene 13605 | Gm13605 | OTTMUSG00000012918 | 22.7kb | Anti-sense | Chr. 2 - 35223694 - 35254164 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab14
Genomic Map