Assay Details
| Gene Symbol: C530042K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043921.1 | - | 1 | 351 | 73 | |
| AK049699.1 | - | 1 | 482 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carbamoyl-phosphate synthetase 1 | Cps1 |
4732433M03Rik, CPS, D1Ucla3 |
-1.8mb | Anti-sense | Chr. 1 - 67122426 - 67231267 on NCBI Build GRCm38 |
| Downstream | predicted gene 28497 | Gm28497 | 431.0kb | Anti-sense | Chr. 1 - 69431135 - 69444090 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C530042K13Rik
Genomic Map