Assay Details
| Gene Symbol: 6030443J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006545.1 | - | 470 | 76 | ||
| AK044020.1 | - | 757 | 76 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | RIKEN cDNA A930003O13 gene | A930003O13Rik | 37.4kb | Anti-sense | Chr. 5 - 22738910 - 22746884 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

6030443J06Rik
Genomic Map