Assay Details
| Gene Symbol: 4122401K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014465.1 | - | 1 | 139 | 98 | |
| AK044899.1 | - | 1 | 144 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 111, member A |
Fam111a |
4632417K18Rik, AW413625 |
-6.0kb | Sense | Chr. 19 - 12573075 - 12589769 on NCBI Build GRCm38 |
| Downstream | predicted gene 4952 | Gm4952 | EG240549 | 2.4kb | Sense | Chr. 19 - 12599984 - 12627616 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4122401K19Rik
Genomic Map