Assay Details
| Gene Symbol: Gm4524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045496.1 | - | 1 - 2 | 65 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transformation related protein 63 | Trp63 |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
-64.4kb | Sense | Chr. 16 - 25683763 - 25892103 on NCBI Build GRCm38 |
| Downstream | claudin 1 | Cldn1 | AI596271 | 396.3kb | Anti-sense | Chr. 16 - 26356645 - 26371839 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm4524
Genomic Map