Assay Details
| Gene Symbol: B230311B06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045786.1 | - | 2 - 3 | 169 | 78 | |
| AW046168.1 | - | 1 - 2 | 78 | |||
| BB309504.2 | - | 2 - 3 | 179 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane channel-like gene family 7 | Tmc7 |
1700030H01Rik, C230064B05, C630024K23Rik |
-10.2kb | Anti-sense | Chr. 7 - 118535841 - 118584790 on NCBI Build GRCm38 |
| Downstream | transmembrane channel-like gene family 5 | Tmc5 |
4932443L08Rik, AU016085 |
473.0 | Sense | Chr. 7 - 118597297 - 118675086 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

B230311B06Rik
Genomic Map