Assay Details
| Gene Symbol: Nacad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046108.1 | - | 1 | 1055 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cerebral cavernous malformation 2 | Ccm2 | BC029157 | -6.3kb | Anti-sense | Chr. 11 - 6546887 - 6596761 on NCBI Build GRCm38 |
| Downstream | transforming growth factor beta regulated gene 4 | Tbrg4 |
2310042P22Rik, AA120735, AA408001, AI527316, Cpr2, R74877, Tb-12, mKIAA0948 |
10.0kb | Sense | Chr. 11 - 6615598 - 6626084 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nacad
Genomic Map