Assay Details
| Gene Symbol: Gm3693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046721.1 | - | 145 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPARC related modular calcium binding 1 | Smoc1 |
2600002F22Rik, AI848508, SRG |
-41.9kb | Sense | Chr. 12 - 81026808 - 81186414 on NCBI Build GRCm38 |
| Downstream | predicted gene 4787 | Gm4787 |
Adam4b, EG214321 |
135.9kb | Anti-sense | Chr. 12 - 81376991 - 81379486 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm3693
Genomic Map