Assay Details
| Gene Symbol: C030017B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046738.1 | - | 1 | 2357 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel tetramerisation domain containing 16 | Kctd16 |
4930434H12Rik, Gm1267 |
-8.8kb | Sense | Chr. 18 - 40258361 - 40531184 on NCBI Build GRCm38 |
| Downstream | PRELI domain containing 2 | Prelid2 |
1700003A01Rik, C330008K14Rik |
1.3mb | Anti-sense | Chr. 18 - 41875696 - 41951194 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030017B01Rik
Genomic Map