Assay Details
| Gene Symbol: Ccdc127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168658.1 | NP_001162129.1 | 4 | 5205 | 89 | |
| GenBank mRNA | AK047833.1 | - | 1 | 857 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 14B | Lrrc14b | AI595366 | -368.0 | Anti-sense | Chr. 13 - 74359582 - 74364000 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 72 | Zfp72 | Zfp74 | 2.9kb | Anti-sense | Chr. 13 - 74368682 - 74390861 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc127
Genomic Map