Assay Details
| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048630.1 | - | 1 - 2 | 78 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930448I06 gene | 4930448I06Rik | -1.5mb | Sense | Chr. 1 - 41167560 - 41181252 on NCBI Build GRCm38 | |
| Downstream |
POU domain, class 3, transcription factor 3 |
Pou3f3 |
Brn-1, Brn1, Otf8, Skin1 |
2.3kb | Anti-sense | Chr. 1 - 42697146 - 42700210 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Pantr1
Genomic Map