Assay Details
| Gene Symbol: C230096K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049070.1 | - | 1 | 1650 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105244755 | LOC105244755 | -62.4kb | Anti-sense | Chr. 5 - 64708210 - 64741650 on NCBI Build GRCm38 | |
| Downstream | toll-like receptor 1 | Tlr1 | 118.9kb | Anti-sense | Chr. 5 - 64924680 - 64933589 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C230096K16Rik
Genomic Map