Assay Details
| Gene Symbol: 4930432N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015295.1 | - | 1 | 1650 | 118 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ER membrane associated RNA degradation | Ermard |
2210404J11Rik, 2410011O22Rik, AI448938 |
-117.6kb | Sense | Chr. 17 - 15041563 - 15090045 on NCBI Build GRCm38 |
| Downstream | predicted gene 5091 | Gm5091 | C330048F19 | 22.2kb | Anti-sense | Chr. 17 - 15231585 - 15240783 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930432N10Rik
Genomic Map