Assay Details
| Gene Symbol: 4933409F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015322.1 | - | 1 - 2 | 409 | 102 | |
| AK016753.1 | - | 1 - 2 | 107 | 102 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY714874.1 | - | 401 | Chr. - - [-] on Build | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700020D12 gene | 1700020D12Rik | -219.4kb | Sense | Chr. 12 - 26572113 - 26613757 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 20187 |
Gm20187 | 255.4kb | Sense | Chr. 12 - 27090237 - 27114995 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4933409F18Rik
Genomic Map