Assay Details
| Gene Symbol: 4930447C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015398.1 | - | 1 | 752 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aryl hydrocarbon receptor nuclear translocator-like 2 | Arntl2 |
4632430A05Rik, BMAL2, CLIF, MOP9, bHLHe6 |
-12.7kb | Anti-sense | Chr. 6 - 146796015 - 146833529 on NCBI Build GRCm38 |
| Downstream | single-pass membrane protein with coiled-coil domains 2 | Smco2 | 1700023A16Rik | 2.9kb | Anti-sense | Chr. 6 - 146850093 - 146871407 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930447C11Rik
Genomic Map