Assay Details
| Gene Symbol: 4930448D08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015414.1 | - | 1 - 2 | 37 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 68 | Ccdc68 | BC046343 | -193.0 | Sense | Chr. 18 - 69924858 - 69969484 on NCBI Build GRCm38 |
| Downstream |
RAB27B, member RAS oncogene family |
Rab27b |
2310021G14Rik, B130064M09Rik |
6.8kb | Anti-sense | Chr. 18 - 69979131 - 70141633 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930448D08Rik
Genomic Map