Assay Details
| Gene Symbol: 4930451A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015436.1 | - | 1 | 452 | 90 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY714982.1 | - | 449 | Chr. - - [-] on Build | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibrous sheath-interacting protein 1 | Fsip1 |
1700012M13Rik, 4933432K11Rik |
-14.2kb | Sense | Chr. 2 - 118130261 - 118256987 on NCBI Build GRCm38 |
| Downstream | G protein-coupled receptor 176 | Gpr176 | Gm1012 | 5.3kb | Sense | Chr. 2 - 118277096 - 118373419 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930451A11Rik
Genomic Map