Assay Details
| Gene Symbol: 4930456J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015477.1 | - | 1 | 1455 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DCN1, defective in cullin neddylation 1, domain containing 3 (S. cerevisiae) |
Dcun1d3 |
1700020A13Rik, BC030335 |
-524.0 | Anti-sense | Chr. 7 - 119853158 - 119896169 on NCBI Build GRCm38 |
| Downstream |
dynein, axonemal, heavy chain 3 |
Dnah3 |
BC051401, Dnahc3 |
24.3kb | Anti-sense | Chr. 7 - 119922671 - 120095675 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930456J16Rik
Genomic Map