Assay Details
| Gene Symbol: 4930470F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015532.1 | - | 1 | 850 | 67 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AI154484.1 | - | Chr. - - [-] on Build | 67 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAB11 family interacting protein 2 (class I) | Rab11fip2 |
4930470G04Rik, A830046J09Rik, AW558126, Nrip11 |
-74.0 | Anti-sense | Chr. 19 - 59760441 - 59943666 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA E330013P04 gene | E330013P04Rik |
9930018I18Rik, AW492152 |
200.0kb | Sense | Chr. 19 - 60144675 - 60162591 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930470F04Rik
Genomic Map