Assay Details
| Gene Symbol: 4930470O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015539.1 | - | 1 - 2 | 185 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC030500 | BC030500 | -911.4kb | Sense | Chr. 8 - 58911598 - 58914298 on NCBI Build GRCm38 | |
| Downstream | aminoadipate aminotransferase | Aadat |
AI875679, Aadt, KATII, Kat2, mKat-2 |
677.2kb | Sense | Chr. 8 - 60505870 - 60545677 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930470O06Rik
Genomic Map