Assay Details
Gene Symbol: 4930473H19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015566.1 | - | 1 - 2 | 36 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | aarF domain containing kinase 1 | Adck1 | 2610005A10Rik | Anti-sense | Chr. 12 - 88360491 - 88461726 on NCBI Build GRCm38 | |
Downstream | neurexin III | Nrxn3 | 280.6kb | Anti-sense | Chr. 12 - 88722865 - 90334935 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
