Assay Details
| Gene Symbol: 4930473H19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015566.1 | - | 2 - 3 | 253 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aarF domain containing kinase 1 | Adck1 | 2610005A10Rik | Anti-sense | Chr. 12 - 88360491 - 88461726 on NCBI Build GRCm38 | |
| Downstream | neurexin III | Nrxn3 | 281.6kb | Anti-sense | Chr. 12 - 88722865 - 90334935 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930473H19Rik
Genomic Map