Assay Details
| Gene Symbol: 4930482G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108044.1 | 2 - 3 | 519 | 99 | ||
| GenBank mRNA | AK015610.1 | - | 2 - 3 | 521 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5 | St6galnac5 |
AI851940, Siat7e |
-165.7kb | Sense | Chr. 3 - 152820709 - 152982207 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700094M23 gene | 1700094M23Rik | 12.7kb | Sense | Chr. 3 - 153162834 - 153183224 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930482G09Rik
Genomic Map