Assay Details
| Gene Symbol: 4930509H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015736.1 | - | 1 | 1152 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine-rich repeats and immunoglobulin-like domains 2 | Lrig2 |
4632419I10Rik, BB096938 |
Sense | Chr. 3 - 104453980 - 104511938 on NCBI Build GRCm38 | |
| Downstream |
solute carrier family 16 (monocarboxylic acid transporters), member 1 |
Slc16a1 |
AL022710, Mct1 |
127.4kb | Anti-sense | Chr. 3 - 104638664 - 104658462 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930509H03Rik
Genomic Map