Assay Details
| Gene Symbol: Gm10306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015743.1 | - | 1 | 351 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tumor suppressor candidate 1 | Tusc1 |
2200001D17Rik, TSG-9 |
-1.2mb | Anti-sense | Chr. 4 - 93334148 - 93335511 on NCBI Build GRCm38 |
| Downstream |
phospholipase A2, activating protein |
Plaa |
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3 |
8.1kb | Anti-sense | Chr. 4 - 94565139 - 94603247 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm10306
Genomic Map