Assay Details
| Gene Symbol: 4930517J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015815.1 | - | 1 | 1357 | 84 | |
| AW488177.1 | - | 1 | 84 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102632357 | LOC102632357 | Sense | Chr. 4 - 44660394 - 44683684 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 5730488B01 gene | 5730488B01Rik | 31.9kb | Sense | Chr. 4 - 44702800 - 44705110 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930517J16Rik
Genomic Map