Assay Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015910.1 | - | 1 | 250 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interleukin 12b | Il12b |
Il-12b, Il-12p40, Il12p40, p40 |
-53.8kb | Anti-sense | Chr. 11 - 44400063 - 44414677 on NCBI Build GRCm38 |
| Downstream | ring finger protein 145 | Rnf145 |
3732413I11Rik, TMRF1 |
48.3kb | Anti-sense | Chr. 11 - 44518680 - 44565520 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ublcp1
Genomic Map