Assay Details
| Gene Symbol: 4930533B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015952.1 | - | 1 | 446 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transformation related protein 73 | Trp73 |
TAp73, Tp73, deltaNp73, p73 |
Anti-sense | Chr. 4 - 154056249 - 154140208 on NCBI Build GRCm38 | |
| Downstream |
WD repeat containing, antisense to Trp73 |
Wrap73 |
2610044M17Rik, 5330425N03Rik, Dd57, Wdr8 |
53.4kb | Sense | Chr. 4 - 154142357 - 154168259 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930533B18Rik
Genomic Map