Assay Details
| Gene Symbol: 4930540A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016005.1 | - | 1 | 353 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39127 |
Gm39127 | -76.3kb | Anti-sense | Chr. 8 - 8722957 - 8755164 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4933430N04 gene | 4933430N04Rik | 293.5kb | Sense | Chr. 8 - 9125412 - 9132288 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930540A11Rik
Genomic Map