Assay Details
| Gene Symbol: 4930544L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016038.1 | - | 1 | 1846 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6391 | Mir6391 |
Gm27914, mmu-mir-6391 |
-71.0kb | Anti-sense | Chr. 14 - 118592369 - 118592472 on NCBI Build GRCm38 |
| Downstream | predicted gene 27198 | Gm27198 | 52.0kb | Anti-sense | Chr. 14 - 118717584 - 118735153 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930544L18Rik
Genomic Map