Assay Details
| Gene Symbol: 4930548F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016066.1 | - | 1 | 1147 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box C1 | Foxc1 |
FREAC3, Fkh1, Mf1, Mf4, ch, fkh-1, frkhda |
-527.0 | Sense | Chr. 13 - 31806646 - 31810635 on NCBI Build GRCm38 |
| Downstream |
GDP-mannose 4, 6-dehydratase |
Gmds |
BC031788, C87208 |
7.1kb | Anti-sense | Chr. 13 - 31819583 - 32338725 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930548F15Rik
Genomic Map